tert-Butyl 2-bromoisobutyrate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23877-12-5
:
Formula:C₆H₁₁BrO₂
MDL Number:MFCD00051908
:223.12 g/mol
Boiling Pt:37°C
EINECS:245-924-7
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together