Order Entry
Italy
ContactUsLinkComponent
Methyl-3,5-dibromobenzoate ≥98%
Irritant
Methyl-3,5-dibromobenzoate ≥98%
Catalog # B20668.06
CAS Number:  
Irritant
Methyl-3,5-dibromobenzoate ≥98%
Catalog # B20668.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:51329-15-8
 :
Formula:C₈H₆Br₂O₂
MDL Number:MFCD00082648
 :293.94 g/mol
Melting Pt:61…64 °C
 :GHS07
Storage Temperature:Ambient