
2,2':6',2''-Terpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1148-79-4
:
Formula:C₁₅H₁₁N₃
MDL Number:MFCD00006213
:233.27 g/mol
Boiling Pt:370°C
EINECS:214-559-5
UN:2811
ADR:6.1,I
:GHS06
Specification Test Results
- Appearance:White to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:91.0 to 95.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together