
3,3-Diphenylpropionitrile ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2286-54-6
:
Formula:C₁₅H₁₃N
MDL Number:MFCD00129747
Boiling Pt:174°C
:207.27 g/mol
EINECS:218-926-0
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 97.0 %
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together