
2-(Trimethylsilyl)pyridine ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13737-04-7
:
Formula:C₈H₁₃NSi
MDL Number:MFCD06797181
:151.28 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %
- Specific gravity (20/20):0.9100 to 0.9300
- Refractive index n20/D:1.4900 to 1.4940
Specification Test Results
Frequently Bought Together