
Diphenyliodonium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:66003-76-7
:
Formula:C₁₃H₁₀F₃IO₃S
MDL Number:MFCD00191356
:430.19 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:177.0 to 183.0 deg-C
Specification Test Results
Frequently Bought Together