Order Entry
Italy
ContactUsLinkComponent
3,4-Dimethylthiobenzamide ≥97%
Irritant
3,4-Dimethylthiobenzamide ≥97%
Catalog # H51793.03
CAS Number:  
Irritant
3,4-Dimethylthiobenzamide ≥97%
Catalog # H51793.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :165.25 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:58952-03-7
Formula:C₉H₁₁NS
MDL Number:MFCD06738326