Order Entry
Italy
ContactUsLinkComponent
4,4'-Thiobisbenzenethiol ≥98.0% (by GC)
Irritant
4,4'-Thiobisbenzenethiol ≥98.0% (by GC)
Catalog # TCIAT1427-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
4,4'-Thiobisbenzenethiol ≥98.0% (by GC)
Catalog # TCIAT1427-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %
  • Melting point:
    113.0 to 119.0 deg-C

Specification Test Results

About this item

 :
 :250.4028 g/mol
Boiling Pt:148°C
 :GHS07
CAS Number:19362-77-7
Formula:C₁₂H₁₀S₃
MDL Number:MFCD00142119