Methyl-2-(methylthio)benzoate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3704-28-7
:
Formula:C₉H₁₀O₂S
MDL Number:MFCD00068039
:182.24 g/mol
Melting Pt:64…66 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together