
Methyl-3-chloro-5-iodobenzoate ≥98%
Catalog # B25444.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
About this item
CAS Number:289039-85-6
:
Formula:C₈H₆ClIO₂
MDL Number:MFCD00673018
:296.49 g/mol
Melting Pt:41…43 °C
:GHS07
Storage Temperature:Ambient