Order Entry
Italy
ContactUsLinkComponent
5-Benzylmercaptotetrazole (BMT, BTT) ≥98.0% (by HPLC, titration analysis)
Flammable
Irritant
5-Benzylmercaptotetrazole (BMT, BTT) ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3020-25G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
5-Benzylmercaptotetrazole (BMT, BTT) ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB3020-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :192.24 g/mol
Melting Pt:133°C
UN:1325
ADR:4.1,III
 :GHS02
 :GHS07
CAS Number:21871-47-6
Formula:C₈H₈N₄S
MDL Number:MFCD00068731