
1,5-Pentamethylene-1H-tetrazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:54-95-5
:
Formula:C₆H₁₀N₄
MDL Number:MFCD00005939
:138.17 g/mol
Merck Index:14,07141
Boiling Pt:194°C
EINECS:200-219-3
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:59.0 to 63.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together