Order Entry
Italy
ContactUsLinkComponent
Cyclopropylmagnesium bromide ∼10% (w/w) ∼0.7 mol/L in THF
Flammable
Corrosive
Irritant
Health
Cyclopropylmagnesium bromide ∼10% (w/w) ∼0.7 mol/L in THF
Catalog # TCIAC2039-100G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Health
Cyclopropylmagnesium bromide ∼10% (w/w) ∼0.7 mol/L in THF
Catalog # TCIAC2039-100G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Concentration (sec Butanol method):
    9.0 to 12.0 w/w%

Specification Test Results

About this item

 :
 :145.28 g/mol
Flash Pt:-17°C
EINECS:472-500-0
UN:3399
ADR:4.3,I
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:23719-80-4
Formula:C₃H₅BrMg
MDL Number:MFCD00672094
Flash Pt:–17°C