



Cyclopropylmagnesium bromide 0.5-0.7 M in THF
Catalog # H26273.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
CAS Number:23719-80-4
:
Formula:C₃H₅BrMg
MDL Number:MFCD00672094
:145.28 g/mol
Density:0.968
Flash Pt:–17 °C (1 °F)
UN:2924
ADR:3,II
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient