Order Entry
Italy
ContactUsLinkComponent
 
 
 
cis-1,2-Cyclohexanedicarboxylic anhydride, Sigma-Aldrich®
  8.20365.1000
 :  Merck
 :  
 
 
cis-1,2-Cyclohexanedicarboxylic anhydride, Sigma-Aldrich®
  8.20365.1000
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Solubility:
    7 g/l (20 °C) (slow decomposition)
  • Melting Point:
    30 - 33 °C
  • Molar Mass:
    154.16 g/mol
  • Boiling Point:
    296 °C (1013 hPa)
  • Vapor Pressure:
    0.01 hPa (20 °C)
  • Flash Point:
    152 °C
  • Density:
    1.23 g/cm3 (20 °C)
  • Ignition Point:
    395 °C DIN 51794

 

 

 :
 :M-Clarity™ quality level = MQ200
 :236-086-3
 :GHS05
 :GHS08
 :Ambient
 :13149-00-3
 :85-42-7
 :C₈H₁₀O₃
 :MFCD00005926