HPLC columns, Hypersil™ MOS and MOS-2 C8
Supplier: Thermo Fisher Scientific
Certificates
About this item
Hypersil™ MOS and MOS-2 columns have a monolayer coverage of C8 alkyl chain chemically bonded onto the silica surface for a reproducible and efficient stationary phase. The MOS-2 is endcapped. Both C8 phases exhibit similar selectivity to ODS, but are less retentive. This can often provide reduced time for analyses, particularly for strongly hydrophobic analytes.
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Dependable and affordable
Frequently Bought Together