Order Entry
Italy
ContactUsLinkComponent
Fleece hat, with Insulatex™ lining, HA10
Fleece hat, with Insulatex™ lining, HA10
Supplier:  Portwest
Certificates
undefined
Fleece hat, with Insulatex™ lining, HA10
Supplier:  Portwest
Specifications
  • Catalog No:
  • PRTWHA10BKR
  • PRTWHA10ORR
  • PRTWHA10YER
  • PRTWHA10NAR
  • Model:
  • HA10
  • HA10
  • HA10
  • HA10
  • Garment application:
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • General Purpose, Industrial
  • Risk area:
  • Cold
  • Cold
  • Cold
  • Cold
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Fabric:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Weight:
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²
  • 280 g/m²