High visibility traffic jacket, 4-in-1, S468
Supplier: Portwest
Certificates
About this item
Breathable jacket made of 300 Denier polyester yarn which features superior abrasion and tear resistance. The PU coating on the inside makes the fabric durable and waterproof. The water- and stain-repellent coating on the outside helps to keep the garment clean. With nylon lining.
- With detachable, reversible bodywarmer
- Two hip pockets with Velcro fastened flaps, inside pocket, mobile phone pocket
- Drawstring hood and hem, knitted storm cuffs
- Taped waterproof seams
- Two-way zip front with studded storm flap
Weight: 190 g/m²
Certifications: EN ISO 20471 (Class 3), EN 343 (3-1)
Specifications
- Catalog No:
- PRTWS468YERS
- PRTWS468YERL
- PRTWS468YERM
- PRTWS468YERXL
- PRTWS468YER4XL
- PRTWS468YERXXL
- PRTWS468ORRM
- PRTWS468ORRS
- PRTWS468ORRXXXL
- PRTWS468YER5XL
- PRTWS468YERXXXL
- PRTWS468ORRL
- PRTWS468YERXS
- PRTWS468ORRXS
- PRTWS468ORRXL
- PRTWS468ORR4XL
- PRTWS468ORRXXL
- Model:
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- S468
- Garment application:
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Outdoor
- Risk area:
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Rain (waterproof), Cold
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn
- 300 Denier polyester yarn