Order Entry
Italy
ContactUsLinkComponent
Flame-retardant rain trousers, high visibility, Tielson 5874
Flame-retardant rain trousers, high visibility, Tielson 5874
Supplier:  SIOEN
Certificates
undefined
Flame-retardant rain trousers, high visibility, Tielson 5874
Supplier:  SIOEN
Specifications
  • Catalog No:
  • SIOE5874/278/L
  • SIOE5874/278/3XL
  • SIOE5874/278/2XL
  • SIOE5874/278/1XL
  • SIOE5874/279/S
  • SIOE5874/279/M
  • SIOE5874/279/L
  • SIOE5874/279/3XL
  • SIOE5874/279/2XL
  • SIOE5874/279/1XL
  • SIOE5874/278/S
  • SIOE5874/278/M
  • Model:
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Tielson 5874
  • Garment application:
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Risk area:
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Rain (waterproof), Flame-retardant, Anti-static
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • 100% polyester fabric with 100% FR PU coating
  • Weight:
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²