Hand cleanser dispensers, Eurospender, 1 Plus
Supplier: BODE CHEMIE
Certificates
About this item
The Eurospender 1 Plus are versatile and user-friendly wall-mounting dispensers. The pump can be quickly exchanged through front removal.
- Robust and reliable metal dispensers which meet high hygiene standards
- Dosing amount adjustable to approx. 0,75 to 1,5 ml per actuation
- Dispensing carried out via the operating lever
Delivery: Including mounting parts, instruction sheet and manual. Accessories and spare parts are available on request. Please contact your local Avantor customer service centre for further information.
Frequently Bought Together