Rotors for MIKRO 220 centrifuge
Supplier: Hettich
About this item
5 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
- Catalog No:
- HETT1015
- 521-1896
- 521-1186
- 521-1187
- 521-1188
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
- ISO 14001 (Cert. Reg. No: 12100/10459604 TMS)
Specifications
Product Family Options
Product Information
- AvailabilityPrice
- HETT1015Fixed angle rotor 1015, 12×15 ml conical tubes, 6000 min-1, 4146 ×g, for use with MIKRO 220 / 220RFixed angle rotor 1015, 12×15 ml conical tubes, 6000 min-1, 4146 ×g, for use with MIKRO 220 / 220RLog in for availability1 * 1 pezzi442,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.HETT1015Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Cert. Reg. No: 12100/10459604 TMS)Low Manufacturing Impact - 521-1896Fixed angle rotor, 6×50 ml conical tubes, 6000 min⁻¹, 4025×g, for use with MIKRO 220 RFixed angle rotor, 6×50 ml conical tubes, 6000 min⁻¹, 4025×g, for use with MIKRO 220 RLog in for availability1 * 1 pezzi665,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.521-1896Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Cert. Reg. No: 12100/10459604 TMS)Low Manufacturing Impact - 521-1186Fixed angle rotor 1158-L, 48×1,5/2,0 ml (2 rows), 14000 min⁻¹, 21255×g, with biocontainment lidFixed angle rotor 1158-L, 48×1,5/2,0 ml (2 rows), 14000 min⁻¹, 21255×g, with biocontainment lidLog in for availability1 * 1 pezzi922,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.521-1186Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Cert. Reg. No: 12100/10459604 TMS)Low Manufacturing Impact - 521-1187Fixed angle rotor 1189-A, 30×1,5/2,0 ml, 14000 min⁻¹, 21255×g, with biocontainment lidFixed angle rotor 1189-A, 30×1,5/2,0 ml, 14000 min⁻¹, 21255×g, with biocontainment lidLog in for availability1 * 1 pezzi645,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.521-1187Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Cert. Reg. No: 12100/10459604 TMS)Low Manufacturing Impact - 521-1188Fixed angle rotor 1195-A, 24×1,5/2,0 ml, 18000 min⁻¹, 31514×g, with biocontainment lidFixed angle rotor 1195-A, 24×1,5/2,0 ml, 18000 min⁻¹, 31514×g, with biocontainment lidLog in for availability1 * 1 pezzi516,00 €Specifications:
Low Manufacturing Impact More
Specifications
Cat. No.521-1188Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001 (Cert. Reg. No: 12100/10459604 TMS)Low Manufacturing Impact