LAL-test, Pyrell, turbidimetric formulation (method C and F, EP), PYROTELL®-T
Certificates
About this item
Pyrotell®-T lysate is used to quantify endotoxin in kinetic turbidimetric tests. Pyrotell-T lysate can be used with the Pyros Kinetix® tube reader and also incubating microplate readers. When used with the Pyros Kinetix tube reader, Pyrotell-T gives a highly economic, flexible and sensitive LAL assay. It can be used for a wide variety of endotoxin tests, ranging from standard water testing to samples requiring high sensitivity, such as intrathecal products and those requiring high dilutions to overcome interference.
Frequently Bought Together