Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Microbiology Test Systems
Endotoxin Tests
LAL- test, standards
LAL- test, standards
LAL- test, standards
Catalog # PB18174
Supplier:
Per processare correttamente l'ordine inserire il n° lotto nel campo 'Commenti'
CAS Number:
Expand image
LAL- test, standards
Catalog # PB18174
Supplier:
Per processare correttamente l'ordine inserire il n° lotto nel campo 'Commenti'
CAS Number:
Specifications
Pk:
1
Description:
Reference Standard Endotoxins (RSE), EP, 10,000 I.U./vial
Specifications
About this item
Product Details
Product Details