LAL- test, water
Certificates
About this item
LAL Reagent Water (LRW) in plastic bottles with screwed cap. It is manufactured in accordance with a validated procedure. This water (LRW) is under strict quality control in accordance with EP and USP and fulfills the following specifications:
- Content of endotoxin: <0,001 EU/ml
- Content of (1—>33)-β-D-glucan: <1,56 pg/ml
Frequently Bought Together