Order Entry
Italy
Orders LinkContactUsLinkComponent
Industrial scales, Explorer®
Industrial scales, Explorer®
Industrial scales, Explorer®
Supplier:  
Certificates
Industrial scales, Explorer®
Industrial scales, Explorer®
0 Options Available Below
Product Details & Documents
Large platform bench scales that incorporate touch screen, advanced weighing and proximity sensor technologies. The easy to use control unit can be mounted on the base, above the base on an optional column, on a wall, or remotely (up to 3 metres distance with an optional cable). The touch screen display, with universal icons and informative prompts, simplifies set-up functions and basic operation. The operator has the added choice to use proximity sensors for hands-free operation for handling sensitive samples. The balances have multiple application modes for basic weighing to SQC applications together with short stabilisation times and optimised performance specifications.
  • Colour touch screen display with viewing angle adjustment in a demountable control unit
  • Advanced weighing technology with stabilisation times of approximately 1 second
  • Icon-driven menu for ease of navigation to required functionality
  • AutoCal™ internal calibration
  • Dual multi-functional proximity sensors for hands-free operation of basic or user selectable functions
  • LIMS compatibility
The weighing platform base (not control/display unit) has IP 54 protection rating.
Product Family Options
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Impact printer with printer cables (611-2571 and 611-2572), EU/UK plugs
    Impact printer with printer cables (611-2571 and 611-2572), EU/UK plugs
    611-2573
    Log in for availability
    1 * 1 pezzi - 800,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Ink ribbon cassette for printer SF40A
    Ink ribbon cassette for printer SF40A
    611-2579
    Log in for availability
    1 * 1 pezzi - 13,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Paper roll for printer SF40A, 57,5 mm width
    Paper roll for printer SF40A, 57,5 mm width
    611-2580
    Log in for availability
    1 * 2 pezzi - 13,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Terminal extension cable (RS422) for Explorer® balances
    Terminal extension cable (RS422) for Explorer® balances
    611-2417
    Log in for availability
    1 * 1 pezzi - 25,20 €