Magnetic stirrer, accuMIX
Supplier: 2MAG
Certificates
About this item
Ultra flat, wear-free magnetic stirrer with stainless steel housing constructed for the independent battery- and stationary mains operation.
- Maintenance- and wear-free inductive drive
- Jerk-free stirring at low speeds
- Two-step power setting for high power with regard to large and viscous stirring amounts
- Reduced power for battery-saving long-term operation
SoftStart for reliable catching/centering and safe accelerating of stirring bar.
Certifications: IP protection class: IP 30
Delivery: Supplied with power supply unit, 12 V car adapter cable, battery charger and 8 rechargeable batteries (AA/Mignon, 2 700 mAh).
Frequently Bought Together