Spectromelt® C 20 grinding and tabletting aid, for X-ray analysis, Supelco®
Catalog # 1.13934.5000
Supplier: Merck
CAS Number:
Specifications
- Pk:5 kg
- Seq:4114
Specifications
Specification Test Results
- Identity (IR-spectrum):passes test
- Al (Aluminium):≤ 0.001
- Ba (Barium):≤ 0.001
- Ca (Calcium):≤ 0.001
- Co (Cobalt):≤ 0.001
- Cr (Chromium):≤ 0.001
- Cu (Copper):≤ 0.001
- Fe (Iron):≤ 0.001
- K (Potassium):≤ 0.001
- Mn (Manganese):≤ 0.001
- Mo (Molybdenum):≤ 0.001
- Ni (Nickel):≤ 0.001
- Pb (Lead):≤ 0.001
- Si (Silicon):≤ 0.005
- Sr (Strontium):≤ 0.001
- Ti (Titanium):≤ 0.001
- V (Vanadium):≤ 0.001
Specification Test Results
About this item
:
:M-Clarity™ quality level = MQ100