Order Entry
Italy
ContactUsLinkComponent
Sodium hydroxide monohydrate, Suprapur®, Supelco®
Sodium hydroxide monohydrate, Suprapur®, Supelco®
Supplier:  Merck
Certificates
Sodium hydroxide monohydrate, Suprapur®, Supelco®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    1100 g/l (20 °C)
  • Molar Mass:
    58.01 g/mol
  • Boiling Point:
    1400 °C
  • pH:
    14 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together