
4-Mercaptobenzoic acid ≥95.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1074-36-8
:
Formula:C₇H₆O₂S
MDL Number:MFCD00016617
:154.1863 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:220.0 to 225.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together