2-Chloro-4-(methylsulphonyl)benzoic acid
Catalog # EHERC11447000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:234.66 g/mol
EINECS:406-520-8
CAS Number:53250-83-2
Formula:C₈H₇ClO₄S
MDL Number:MFCD00216496