Order Entry
Italy
ContactUsLinkComponent
4'-Formylbenzo-18-crown-6 (4'-formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98.0% (by GC)
Irritant
4'-Formylbenzo-18-crown-6 (4'-formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98.0% (by GC)
Catalog # TCIAF0451-500MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
4'-Formylbenzo-18-crown-6 (4'-formylbenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥98.0% (by GC)
Catalog # TCIAF0451-500MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :340.37 g/mol
 :GHS07
CAS Number:60835-74-7
Formula:C₁₇H₂₄O₇
MDL Number:MFCD00388882