

Tetrabutylammonium fluoride 70-75% in water
Supplier: TCI
Certificates
About this item
CAS Number:429-41-4
:
Formula:C₁₆H₃₆FN
MDL Number:MFCD00011747
Merck Index:14,09187
ADR:8,III
:261.47 g/mol
EINECS:207-057-2
UN:1760
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Concentration (Nonaqueous Titration):70.0 to 75.0 w/w%
Specification Test Results
Frequently Bought Together