
1-Methylpyridinium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:7680-73-1
:
Formula:C₆H₈ClN
MDL Number:MFCD00137532
EINECS:231-658-9
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:138.0 to 144.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together