
Tetrabutylammonium perrhenate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:16385-59-4
:
Formula:C₁₆H₃₆NO₄Re
MDL Number:MFCD00012037
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together