Order Entry
Italy
ContactUsLinkComponent
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0% (by GC)
Irritant
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0% (by GC)
Catalog # TCIAC1544-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,4,9,12-Tetraoxadispiro[4.2.4.2]tetradecan ≥99.0% (by GC)
Catalog # TCIAC1544-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    77.0 to 81.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :200.23 g/mol
 :GHS07
CAS Number:183-97-1
Formula:C₁₀H₁₆O₄
MDL Number:MFCD00010851