3,3-Dimethyl-1,5-dioxaspiro[5.5]undecan-9-one ≥98.0% (by GC)
: TCI
:69225-59-8
:
:C₁₁H₁₈O₃
:MFCD00006652
:198.26 g/mol
:273-918-4
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:49.0 to 53.0 deg-C
Frequently Bought Together