Order Entry
Italy
ContactUsLinkComponent
Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid (from Sc2O3) standard for AAS and ICP
Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid (from Sc2O3) standard for AAS and ICP
  PERCN9300148
 :  
Scandium standard solution, 1,000 mg/L Sc in dil. nitric acid (from Sc2O3) standard for AAS and ICP
  PERCN9300148
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Scandium
  • Symbol:
    Sc