Order Entry
Italy
ContactUsLinkComponent
Ethylamine ∼10% ∼2 mol/L in THF
Flammable
Corrosive
Irritant
Health
Ethylamine ∼10% ∼2 mol/L in THF
Catalog # TCIAE0842-100ML
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
Irritant
Health
Ethylamine ∼10% ∼2 mol/L in THF
Catalog # TCIAE0842-100ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Concentration (Neutralization titration):
    10.0 to 13.0 w/w%

Specification Test Results

About this item

 :
 :45.08 g/mol
Merck Index:14,03762
Storage Temperature:Refrigerator
EINECS:200-834-7
UN:2924
ADR:3,II
 :GHS02
 :GHS05
 :GHS07
 :GHS08
CAS Number:75-04-7
Formula:C₂H₇N
MDL Number:MFCD00008160