Order Entry
Italy
ContactUsLinkComponent
N-Alkyl-N,N-dimethyl-N-benzylammonium chloride for synthesis, Sigma-Aldrich®
Corrosive
Irritant
Environment
N-Alkyl-N,N-dimethyl-N-benzylammonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.14363.0500
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
Environment
N-Alkyl-N,N-dimethyl-N-benzylammonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.14363.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (argentometric):
    ≥ 98.0 %
  • Melting range (upper value):
    ≤ 73 °C
  • Identity (IR):
    passes test
  • Flash Point:
    100 °C
  • pH:
    5 - 8 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    250 °C Dust

Specification Test Results

About this item

 :
Melting Pt:73 °C
EINECS:287-089-1
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:1759
 :GHS05
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:85409-22-9
Formula:(C₆H₅CH₂)(CH₃)₂N(C₁₂⁻C₁₄Alkyl)⁺Cl⁻