Di-tert-butylmethylphosphonium tetraphenylborate ≥98.0% (by titrimetric analysis)
Catalog # TCIAD3330-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Iodometric Titration):min. 98.0 %
Specification Test Results
About this item
:
CAS Number:853073-44-6
Formula:C₃₃H₄₂BP
MDL Number:MFCD11869304