Supelco® Organic Reference Standard, Methyl cis-5,8,11,14-eicosatetraenoate solution
Catalog # 47572-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:10 mg/ml in heptane
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:2566-89-4
- Description:Methyl cis-5,8,11,14-eicosatetraenoate solution
- Storage conditions:2…8 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08, GHS07, GHS09
Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331