
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:69891-92-5
:
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00012000
:457.36 g/mol
EINECS:274-189-5
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Related Information
Frequently Bought Together