Order Entry
Italy
ContactUsLinkComponent
Safety shoes, clogs
Safety shoes, clogs
Catalog # 114-3893
Supplier:  Honeywell Safety
undefined
Safety shoes, clogs
Catalog # 114-3893
Supplier:  Honeywell Safety
Specifications
  • Shoe size:
    36
  • Colour:
    Black
  • Pk:
    1 PAIR
Specifications
About this item
Safety clogs with heel straps made from washable microfibre.
  • Textile lining
  • Mono-density PU sole
  • Protective steel toecap (200 J)
The shoes are recommended for people working in laboratories, food industries and other areas where hygiene is required.
Colour: Black or white
Width: 11
Certifications: EN ISO 20345; SB, A, E, SRC.