Capillary columns, TraceGOLD™ TG-1701MS
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns feature a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns.
- Mid-polarity phase, 14% cyanopropylphenyl, 86% dimethyl polysiloxane
- Fully characterised for long-term reproducibility, column-to-column consistency and low bleed
- Optimal for confirmation analysis
- Equivalent to USP G46 phase
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together