Order Entry
Italy
ContactUsLinkComponent
 
DNA/RNA isolation, viral DNA/RNA 96 kit, Mag-Bind®
 
undefined
DNA/RNA isolation, viral DNA/RNA 96 kit, Mag-Bind®

 

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Viral RNA
    Viral DNA
  • Sample type:
    Swabs
    Blood
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Sample size:
    50 - 200 μl

 

Frequently Bought Together