
2-(Hydroxymethyl)-15-crown-5 ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:75507-25-4
:
Formula:C₁₁H₂₂O₆
MDL Number:MFCD00192215
:250.29 g/mol
Boiling Pt:107°C
EINECS:278-222-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Related Information
Frequently Bought Together