Order Entry
Italy
ContactUsLinkComponent
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Supplier:  TCI
Certificates
undefined
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Supplier:  TCI
Specification Test Results
  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Lambda max.:
    531.0 to 535.0 nm (MeOH)
  • Absorbance (E1%1cm):
    min. 750 (MeOH, 531.0 to 535.0 nm)