Order Entry
Italy
ContactUsLinkComponent
2-(1-Ethylpropyl)pyridine ≥95.0% (by GC)
2-(1-Ethylpropyl)pyridine ≥95.0% (by GC)
Catalog # TCIAP0969-25ML
Supplier:  TCI
CAS Number:  
undefined
2-(1-Ethylpropyl)pyridine ≥95.0% (by GC)
Catalog # TCIAP0969-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :149.24 g/mol
EINECS:230-998-5
CAS Number:7399-50-0
Formula:C₁₀H₁₅N
MDL Number:MFCD00014631