Order Entry
Italy
ContactUsLinkComponent
Sodium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Oxidizing
Irritant
Sodium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Oxidizing
Irritant
Sodium nitrate ≥99.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    ≥99.0%
  • For Laboratory,Research,or Manufacturing Use:
    5.5 - 8.3
  • ACS - Assay (NaNO₃):
    ≥99.0 %
  • ACS - pH of 5% Solution at 25°C:
    5.5 - 8.3
  • Insoluble Matter:
    ≤0.005 %
  • Chloride (Cl):
    ≤0.001 %
  • ACS - Iodate (IO₃):
    ≤5 ppm
  • ACS- Nitrite (NO₂):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤5 ppm
  • ACS - Sulfate (SO₄):
    ≤0.003 %
  • Calcium (Ca):
    ≤0.005 %
  • ACS - Magnesium (Mg):
    ≤0.002 %
  • ACS - Heavy Metals (as Pb):
    ≤5 ppm
  • ACS - Iron (Fe):
    ≤3 ppm

Specification Test Results

Related Information

Frequently Bought Together