


1-Ethoxy-2,2,2-trifluoroethanol ≥80.0% (by GC) (contains ca. 10% Ethanol)
Catalog # TCIAT0791-10G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 80.0 %
Specification Test Results
About this item
:
:144.09 g/mol
Flash Pt:39°C
EINECS:207-087-6
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:433-27-2
Formula:C₄H₇F₃O₂
MDL Number:MFCD00000441